accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58028-76-5

IUPAC Name: 2-(morpholin-4-yl)benzaldehyde
Synonyms: 2-(morpholin-4-yl)benzaldehyde
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: GTTAEWVBVHSDLX-UHFFFAOYSA-N
SMILES: O=CC1=CC=CC=C1N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results