accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSO

CAS RN 58066-86-7

IUPAC Name: 1-{[(furan-2-yl)methyl]sulfanyl}propan-2-one
Synonyms: 1-[(furan-2-ylmethyl)sulfanyl]propan-2-one
Molecular Weight (g/mol): 170.23
Molecular Formula: C8H10O2S
InChi Key: WWWHLSMCOBIZNV-UHFFFAOYSA-N
SMILES: CC(=O)CSCC1=CC=CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results