accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5807-30-7

OHOClCl

CAS RN 5807-30-7

IUPAC Name: 2-(3,4-dichlorophenyl)acetic acid
Synonyms: (3,4-dichlorophenyl)acetic acid
Molecular Weight (g/mol): 205.03
Molecular Formula: C8H6Cl2O2
InChi Key: ZOUPGSMSNQLUNW-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC=C(Cl)C(Cl)=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)