accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 581-40-8

IUPAC Name: 2,3-dimethylnaphthalene
Synonyms: 2,3-dimethylnaphthalene guajen
Molecular Weight (g/mol): 156.23
Molecular Formula: C12H12
InChi Key: WWGUMAYGTYQSGA-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)