accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5813-64-9

IUPAC Name: 2,2-dimethylpropan-1-amine
Synonyms: 2,2-dimethyl-1-propylamin
Molecular Weight (g/mol): 87.17
Molecular Formula: C5H13N
InChi Key: XDIAMRVROCPPBK-UHFFFAOYSA-N
SMILES: CC(C)(C)CN
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results