accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 583-39-1

IUPAC Name: 2,3-dihydro-1H-1,3-benzodiazole-2-thione
Synonyms: 1,3-dihydro-1,3-benzodiazole-2-thione
Molecular Weight (g/mol): 150.2
Molecular Formula: C7H6N2S
InChi Key: YHMYGUUIMTVXNW-UHFFFAOYSA-N
SMILES: S=C1NC2=CC=CC=C2N1
Molecular Weight (g/mol) 
  • (4)
  • (8)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results