accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 583-53-9

IUPAC Name: 1,2-dibromobenzene
Synonyms: 1,2-dibromobenzene O-dibromobenzene
Molecular Weight (g/mol): 235.91
Molecular Formula: C6H4Br2
InChi Key: WQONPSCCEXUXTQ-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)