accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH2Br

CAS RN 583-68-6

IUPAC Name: 2-bromo-4-methylaniline
Synonyms: benzenamine, 2-bromo-4-methyl-
Molecular Weight (g/mol): 186.05
Molecular Formula: C7H8BrN
InChi Key: UVRRJILIXQAAFK-UHFFFAOYSA-N
SMILES: CC1=CC=C(N)C(Br)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)