accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3BrH3C

CAS RN 583-70-0

IUPAC Name: 1-bromo-2,4-dimethylbenzene
Synonyms: benzene, 1-bromo-2,4-dimethyl-
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: YSFLQVNTBBUKEA-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C(C)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)