accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58306-30-2

IUPAC Name: methyl N-{[(methoxycarbonyl)imino]({[2-(2-methoxyacetamido)-4-(phenylsulfanyl)phenyl]amino})methyl}carbamate
Synonyms: methyl N-{[(methoxycarbonyl)imino]({[2-(2-methoxyacetamido)-4-(phenylsulfanyl)phenyl]amino})methyl}carbamate
Molecular Weight (g/mol): 446.48
Molecular Formula: C20H22N4O6S
InChi Key: HMCCXLBXIJMERM-UHFFFAOYSA-N
SMILES: COCC(=O)NC1=CC(SC2=CC=CC=C2)=CC=C1NC(NC(=O)OC)=NC(=O)OC
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)