accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHN

CAS RN 58333-75-8

IUPAC Name: 4-(2-methoxyphenyl)piperidine
Synonyms: 4-(2-methoxyphenyl)piperidine
Molecular Weight (g/mol): 191.27
Molecular Formula: C12H17NO
InChi Key: SRAVSVBVHDLLPO-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results