accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58347-49-2

NNNCl

CAS RN 58347-49-2

IUPAC Name: 7-chloropyrazolo[1,5-a]pyrimidine
Synonyms: 7-chloropyrazolo[1,5-a]pyrimidine
Molecular Weight (g/mol): 153.57
Molecular Formula: C6H4ClN3
InChi Key: YCZQHXPIKQHABJ-UHFFFAOYSA-N
SMILES: ClC1=CC=NC2=CC=NN12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results