CAS RN 58366-64-6
IUPAC Name:
hydrogen 6-amino-5-methyl-1,2-dihydropyrimidin-2-one chloride
Synonyms:
hydrogen 5-methylcytosine chloride
Molecular Weight (g/mol):
161.59
Molecular Formula:
C5H8ClN3O
InChi Key:
ANWMULVRPAUPJT-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC1=C(N)NC(=O)N=C1
1
–
3
of
3
results
1
5-Methyl Cytosine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
5-Methyl Cytosine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.