accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 584-74-7

H3COOF

CAS RN 584-74-7

IUPAC Name: ethyl 2-(2-fluorophenyl)acetate
Synonyms: ethyl 2-(2-fluorophenyl)acetate
Molecular Weight (g/mol): 182.19
Molecular Formula: C10H11FO2
InChi Key: OFWMNCYZASDVSS-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results