accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 584-94-1

H3CCH3CH3CH3

CAS RN 584-94-1

IUPAC Name: 2,3-dimethylhexane
Synonyms: 2,3-dimethylhexane
Molecular Weight (g/mol): 114.23
Molecular Formula: C8H18
InChi Key: JXPOLSKBTUYKJB-UHFFFAOYNA-N
SMILES: CCCC(C)C(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results