CAS RN 58417-15-5
IUPAC Name:
1-(difluoromethoxy)-4-isocyanatobenzene
Synonyms:
1-(difluoromethoxy)-4-isocyanatobenzene
Molecular Weight (g/mol):
185.13
Molecular Formula:
C8H5F2NO2
InChi Key:
XLUSNOUGZQJPRN-UHFFFAOYSA-N
SMILES:
FC(F)OC1=CC=C(C=C1)N=C=O
1
–
1
of
1
results
1
4-(Difluoromethoxy)phenyl isocyanate, 99%, Thermo Scientific™
CAS: 58417-15-5 Molecular Formula: C8H5F2NO2 Molecular Weight (g/mol): 185.13 MDL Number: MFCD00673056 InChI Key: XLUSNOUGZQJPRN-UHFFFAOYSA-N Synonym: 4-difluoromethoxy phenyl isocyanate,1-difluoromethoxy-4-isocyanatobenzene,4-difluoromethoxy benzenisocyanate,acmc-1b0ih,4-difluoromethoxyphenyl isocyanate,4-difluoromethoxy-phenyl isocyanate,4-difluoromethoxy phenyl isosyanate,1-difluoromethoxy-4-isocyanato-benzene,4-difluoromethoxy phenylisocyanate,benzene,1-difluoromethoxy-4-isocyanato PubChem CID: 2733371 IUPAC Name: 1-(difluoromethoxy)-4-isocyanatobenzene SMILES: FC(F)OC1=CC=C(C=C1)N=C=O