accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

SNOOClSOOCl

CAS RN 58457-24-2

IUPAC Name: 5-chloro-4-nitrothiophene-2-sulfonyl chloride
Synonyms: 5-chloro-4-nitrothiophene-2-sulfonyl chloride
Molecular Weight (g/mol): 262.08
Molecular Formula: C4HCl2NO4S2
InChi Key: SGHAHFHTASPNKN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)SC(=C1)S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results