accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2BrFFF

CAS RN 58458-10-9

IUPAC Name: 2-bromo-3-(trifluoromethyl)aniline
Synonyms: 2-bromo-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 240.02
Molecular Formula: C7H5BrF3N
InChi Key: XIUJCAJFRWDEKE-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1Br)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results