accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58477-85-3

OH(S)(R)OHOHNH2CONHCH2

CAS RN 58477-85-3

IUPAC Name: (2R,3S)-2,3-dihydroxy-N,N'-bis(prop-2-en-1-yl)butanediamide
Synonyms: (2R,3S)-2,3-dihydroxy-N,N'-bis(prop-2-en-1-yl)succinamide
Molecular Weight (g/mol): 228.25
Molecular Formula: C10H16N2O4
InChi Key: ZRKLEAHGBNDKHM-OCAPTIKFSA-N
SMILES: O[C@@H]([C@@H](O)C(=O)NCC=C)C(=O)NCC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results