accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 585-29-5

CH3NH3CH3COOH

CAS RN 585-29-5

IUPAC Name: formic acid; triethylamine
Synonyms: formic acid; triethylamine
Molecular Weight (g/mol): 147.22
Molecular Formula: C7H17NO2
InChi Key: PTMFUWGXPRYYMC-UHFFFAOYSA-N
SMILES: OC=O.CCN(CC)CC
Quantity 
  • (3)
  • (2)
For Use With (Equipment) 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results