accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 585-70-6

IUPAC Name: 5-bromofuran-2-carboxylic acid
Synonyms: 5-bromofuran-2-carboxylic acid
Molecular Weight (g/mol): 190.98
Molecular Formula: C5H3BrO3
InChi Key: YVTQHZDUDUCGRD-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(Br)O1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results