accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CBr

CAS RN 585-71-7

IUPAC Name: (1-bromoethyl)benzene
Synonyms: (1-bromoethyl)benzene
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: CRRUGYDDEMGVDY-UHFFFAOYNA-N
SMILES: CC(Br)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)