accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 585-88-6

IUPAC Name: 4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexane-1,2,3,5,6-pentol
Synonyms: maltitol
Molecular Weight (g/mol): 344.31
Molecular Formula: C12H24O11
InChi Key: VQHSOMBJVWLPSR-UHFFFAOYNA-N
SMILES: OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)CO
Molecular Weight (g/mol) 
  • (4)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)