accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONHNHOCH3

CAS RN 58537-94-3

IUPAC Name: 1,7-dimethyl heptanediimidate
Synonyms: dimethyl pimelimidate
Molecular Weight (g/mol): 186.26
Molecular Formula: C9H18N2O2
InChi Key: LRPQMNYCTSPGCX-UHFFFAOYSA-N
SMILES: COC(=N)CCCCCC(=N)OC
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)