accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 586-39-0

ONOCH2

CAS RN 586-39-0

IUPAC Name: 1-ethenyl-3-nitrobenzene
Synonyms: (3-nitrophenyl)ethene
Molecular Weight (g/mol): 149.15
Molecular Formula: C8H7NO2
InChi Key: SYZVQXIUVGKCBJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC(C=C)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results