accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 587-33-7

(S)ONH2OHHO

CAS RN 587-33-7

IUPAC Name: (3R)-2-amino-3-hydroxy-3-phenylpropanoic acid
Synonyms: (3R)-2-amino-3-hydroxy-3-phenylpropanoic acid
Molecular Weight (g/mol): 181.19
Molecular Formula: C9H11NO3
InChi Key: VHVGNTVUSQUXPS-BRFYHDHCSA-N
SMILES: NC([C@H](O)C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)