CAS RN 58777-87-0
IUPAC Name:
1-(4-methoxyphenyl)-2-(methylamino)ethan-1-ol
Synonyms:
1-(4-methoxyphenyl)-2-(methylamino)ethanol
Molecular Weight (g/mol):
181.24
Molecular Formula:
C10H15NO2
InChi Key:
IWLPVZQFUJSAKT-UHFFFAOYNA-N
SMILES:
CNCC(O)C1=CC=C(OC)C=C1
1
–
1
of
1
results
1
1-(4-Methoxyphenyl)-2-(methylamino)ethanol, 97%, Thermo Scientific™
CAS: 58777-87-0 Molecular Formula: C10H15NO2 Molecular Weight (g/mol): 181.235 MDL Number: MFCD08060969 InChI Key: IWLPVZQFUJSAKT-UHFFFAOYSA-N Synonym: 1-4-methoxyphenyl-2-methylamino ethanol,1-4-methoxy-phenyl-2-methylamino-ethanol,1-4-methoxyphenyl-2-methylamino ethan-1-ol,acmc-20apag,1-4-methoxyphenyl-2-methylaminoethanol,benzenemethanol, a-aminomethyl-4-methoxy,1-4-methoxyphenyl-2-methylamino-1-ethanol PubChem CID: 14121158 IUPAC Name: 1-(4-methoxyphenyl)-2-(methylamino)ethanol SMILES: CNCC(C1=CC=C(C=C1)OC)O