CAS RN 587871-26-9
IUPAC Name:
2-(morpholin-4-yl)-6-(thianthren-1-yl)-4H-pyran-4-one
Synonyms:
2-(morpholin-4-yl)-6-(thianthren-1-yl)pyran-4-one
Molecular Weight (g/mol):
395.49
Molecular Formula:
C21H17NO3S2
InChi Key:
XRKYMMUGXMWDAO-UHFFFAOYSA-N
SMILES:
O=C1C=C(OC(=C1)C1=CC=CC2=C1SC1=CC=CC=C1S2)N1CCOCC1
1
–
2
of
2
results
1
KU 55933, TRC
CAS: 587871-26-9 Molecular Formula: C21H17NO3S2 Molecular Weight (g/mol): 395.49 Synonym: KU 55933,ATM Kinase Inhibitor,2-Thianthren-1-yl-6-(morpholin-4-yl)-4H-pyran-4-one SMILES: O=C1C=C(OC(=C1)c2cccc3Sc4ccccc4Sc23)N5CCOCC5