CAS RN 58812-37-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Toosendanin, TRC
CAS: 58812-37-6 Molecular Formula: C30 H38 O11 Molecular Weight (g/mol): 574.62 Synonym: [C(R),1α,3α,4β,5α,7α,12α,13α,14β,15β,17α]-3,12-Bis(acetyloxy)-14,15:21,23-diepoxy-1,7,19-trihydroxy-4,8-dimethyl-11-oxo-24-Norchola-20,22-diene-4-carboxaldehyde Cyclic 4,19-Hemiacetal,14,15-Epoxy-4,10-(methanoxymethano)-1H-cyclopenta[a]phenanthrene, 24-Norchola-20,22-diene-4-carboxaldehyde Deriv.,28-Deacetylsendanin,Chuanliansu SMILES: CC(=O)O[C@@H]1C[C@H](O)[C@@]23CO[C@@H](O)[C@]1(C)[C@@H]2C[C@@H](O)[C@]4(C)[C@@H]3C(=O)[C@H](OC(=O)C)[C@@]5(C)[C@@H](C[C@H]6O[C@@]456)c7cocc7