accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58871-11-7

OH(R)O(R)HO(S)O

CAS RN 58871-11-7

IUPAC Name: (2R,3S,4R)-3-(benzyloxy)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-4-ol
Synonyms: (2R,3S,4R)-3-(benzyloxy)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-4-ol
Molecular Weight (g/mol): 236.27
Molecular Formula: C13H16O4
InChi Key: OIWQZGBSQDJVJN-UPJWGTAASA-N
SMILES: OC[C@H]1OC=C[C@@H](O)[C@@H]1OCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results