accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58871-20-8

IUPAC Name: 2,2-dimethyl-2H,4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl benzoate
Synonyms: 2,2-dimethyl-4H,4aH,8H,8aH-pyrano[3,2-d][1,3]dioxin-8-yl benzoate
Molecular Weight (g/mol): 290.32
Molecular Formula: C16H18O5
InChi Key: RADJUQJHMUFUJI-UHFFFAOYNA-N
SMILES: CC1(C)OCC2OC=CC(OC(=O)C3=CC=CC=C3)C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results