CAS RN 58880-19-6
IUPAC Name:
(2E,4E)-7-[4-(dimethylamino)phenyl]-N-hydroxy-4,6-dimethyl-7-oxohepta-2,4-dienamide
Synonyms:
trichostatin A
Molecular Weight (g/mol):
302.37
Molecular Formula:
C17H22N2O3
InChi Key:
RTKIYFITIVXBLE-WKWSCTOINA-N
SMILES:
CC(\C=C(/C)\C=C\C(=O)NO)C(=O)C1=CC=C(C=C1)N(C)C
1
–
2
of
2
results
2
STEMCELL Technologies Trichostatin A
Epigenetic modifier; Inhibits histone deacetylase (HDAC)1 and HDAC6