accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 589-29-7

HOOH

CAS RN 589-29-7

IUPAC Name: [4-(hydroxymethyl)phenyl]methanol
Synonyms: 1,4-benzenedimethanol
Molecular Weight (g/mol): 138.17
Molecular Formula: C8H10O2
InChi Key: BWVAOONFBYYRHY-UHFFFAOYSA-N
SMILES: OCC1=CC=C(CO)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)