accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3HNO

CAS RN 5894-65-5

IUPAC Name: N-tert-butylbenzamide
Synonyms: N-tert-butylbenzamide
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: HYWWTHOGCJXTRI-UHFFFAOYSA-N
SMILES: CC(C)(C)NC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results