accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58944-73-3

IUPAC Name: (2S,5S)-2,5-diamino-6-[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]hexanoic acid
Synonyms: sinefungin compound 57926 antibiotic 32232 RP antibiotic A 9145 adenosylornithine sinefungine sinefungina sinefunginum
Molecular Weight (g/mol): 381.39
Molecular Formula: C15H23N7O5
InChi Key: LMXOHSDXUQEUSF-YECHIGJVSA-N
SMILES: N[C@@H](CC[C@H](N)C(O)=O)C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C(N)N=CN=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results