accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SH2NCH3

CAS RN 58952-03-7

IUPAC Name: 3,4-dimethylbenzene-1-carbothioamide
Synonyms: 3,4-dimethylbenzenecarbothioamide
Molecular Weight (g/mol): 165.25
Molecular Formula: C9H11NS
InChi Key: MGSZKKVFYBQMPF-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C(C=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results