accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 58960-00-2

IUPAC Name: 2-methyl-3,4-dihydro-2H-1,4-benzothiazine
Synonyms: 2-methyl-3,4-dihydro-2H-1,4-benzothiazine
Molecular Weight (g/mol): 165.25
Molecular Formula: C9H11NS
InChi Key: QZSCUJZRZIIVBM-UHFFFAOYNA-N
SMILES: CC1CNC2=CC=CC=C2S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results