accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59-06-3

IUPAC Name: methyl 4-acetamido-2-ethoxybenzoate
Synonyms: ethopabate
Molecular Weight (g/mol): 237.26
Molecular Formula: C12H15NO4
InChi Key: GOVWOKSKFSBNGD-UHFFFAOYSA-N
SMILES: CCOC1=CC(NC(C)=O)=CC=C1C(=O)OC
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results