accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59-14-3

IUPAC Name: 5-bromo-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: bromodeoxyuridine
Molecular Weight (g/mol): 307.1
Molecular Formula: C9H11BrN2O5
InChi Key: WOVKYSAHUYNSMH-UHFFFAOYNA-N
SMILES: OCC1OC(CC1O)N1C=C(Br)C(=O)NC1=O
Quantity 
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (13)
Physical Form 
  • (4)
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (4)
  • (3)
  • (1)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)
  • (1)
  • (3)
  • (3)
  • (13)
  • (4)
  • (4)
  • (2)
  • (3)
  • (4)