accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59-50-7

IUPAC Name: 4-chloro-3-methylphenol
Synonyms: baktol parol clorocresol baktolan parmetol chlorocresolum rasen-anicon chlorocresolo clorocresolo chlorcresolum
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: CFKMVGJGLGKFKI-UHFFFAOYSA-N
SMILES: CC1=CC(O)=CC=C1Cl
Molecular Weight (g/mol) 
  • (8)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (8)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)