accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 59-66-5

IUPAC Name: N-(5-sulfamoyl-1,3,4-thiadiazol-2-yl)acetamide
Synonyms: acetazolamide nephramid natrionex atenezol acetamox storzolamide cidamex ak-zol nephramide diluran
Molecular Weight (g/mol): 222.24
Molecular Formula: C4H6N4O3S2
InChi Key: BZKPWHYZMXOIDC-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NN=C(S1)S(N)(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results