accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NNH

CAS RN 59-98-3

IUPAC Name: 2-benzyl-4,5-dihydro-1H-imidazole
Synonyms: tolazoline tolazoline preparation divascol tolazolina benzolin dilatol asi benzolin (vasodilator) pridazole 2-benzyl-4,5-imidazoline priscoline
Molecular Weight (g/mol): 160.22
Molecular Formula: C10H12N2
InChi Key: JIVZKJJQOZQXQB-UHFFFAOYSA-N
SMILES: C(C1=NCCN1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results