accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 590-97-6

CH3OOBr

CAS RN 590-97-6

IUPAC Name: bromomethyl acetate
Synonyms: bromomethyl acetate
Molecular Weight (g/mol): 152.98
Molecular Formula: C3H5BrO2
InChi Key: NHYXMAKLBXBVEO-UHFFFAOYSA-N
SMILES: CC(=O)OCBr
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)