accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNBr

CAS RN 590417-55-3

IUPAC Name: 4-bromo-1-methyl-1H-indole
Synonyms: 4-bromo-1-methylindole
Molecular Weight (g/mol): 210.07
Molecular Formula: C9H8BrN
InChi Key: JZOSXTYDJPHXQD-UHFFFAOYSA-N
SMILES: CN1C=CC2=C(Br)C=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results