accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNBrN

CAS RN 590417-94-0

IUPAC Name: 6-bromo-1-methyl-1H-indazole
Synonyms: 6-bromo-1-methylindazole
Molecular Weight (g/mol): 211.06
Molecular Formula: C8H7BrN2
InChi Key: SVVSOIGNROPKMS-UHFFFAOYSA-N
SMILES: CN1N=CC2=CC=C(Br)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results