accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 591-28-6

IUPAC Name: 4-sulfanylidene-1,2,3,4-tetrahydropyrimidin-2-one
Synonyms: 4-sulfanylidene-1,3-dihydropyrimidin-2-one
Molecular Weight (g/mol): 128.15
Molecular Formula: C4H4N2OS
InChi Key: OVONXEQGWXGFJD-UHFFFAOYSA-N
SMILES: O=C1NC=CC(=S)N1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)