accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHClCl

CAS RN 591-35-5

IUPAC Name: 3,5-dichlorophenol
Synonyms: 3,5-dichlorophenol
Molecular Weight (g/mol): 163
Molecular Formula: C6H4Cl2O
InChi Key: VPOMSPZBQMDLTM-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results