accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 591-76-4

IUPAC Name: 2-methylhexane
Synonyms: 2-methylhexane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: GXDHCNNESPLIKD-UHFFFAOYSA-N
SMILES: CCCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results