accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH2

CAS RN 591-87-7

IUPAC Name: prop-2-en-1-yl acetate
Synonyms: allyl acetate
Molecular Weight (g/mol): 100.12
Molecular Formula: C5H8O2
InChi Key: FWZUNOYOVVKUNF-UHFFFAOYSA-N
SMILES: CC(=O)OCC=C
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)