CAS RN 591-93-5
CAS RN 591-93-5
IUPAC Name:
penta-1,4-diene
Synonyms:
1,4-pentadiene
Molecular Weight (g/mol):
68.12
Molecular Formula:
C5H8
InChi Key:
QYZLKGVUSQXAMU-UHFFFAOYSA-N
SMILES:
C=CCC=C
1
–
1
of
1
results
1
1,4-Pentadiene, 99%
CAS: 591-93-5 Molecular Formula: C5H8 Molecular Weight (g/mol): 68.11 MDL Number: MFCD00008665 InChI Key: QYZLKGVUSQXAMU-UHFFFAOYSA-N Synonym: 1,4-pentadiene,unii-a5if861oq5,pentadiene-1,4,allylethylene,penta-1,4-dien-1-yl,penta-1,4-dien-3-yl,ch2=chch2ch=ch2,acmc-1at58 PubChem CID: 11587 IUPAC Name: penta-1,4-diene SMILES: C=CCC=C